Assay Details
| Gene Symbol: drk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057510.4 | NP_476858.1 | 3 - 4 | 524 | 100 | |
| NM_165994.2 | NP_725302.1 | 3 - 4 | 455 | 100 | ||
| NM_165995.2 | NP_725303.1 | 3 - 4 | 426 | 100 | ||
| NM_165996.2 | NP_725304.1 | 2 - 3 | 575 | 100 | ||
| NM_165998.2 | NP_725306.1 | 3 - 4 | 357 | 100 | ||
| GenBank mRNA | AY061142.1 | - | 3 - 4 | 499 | 100 | |
| L12446.1 | - | 3 - 4 | 411 | 100 | ||
More Information
Set Membership: |
|

drk
Genomic Map