Assay Details
| Gene Symbol: TpnC73F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300162.1 | NP_001287091.1 | 2 - 3 | 331 | 73 | |
| NM_079398.4 | NP_524122.2 | 2 - 3 | 234 | 73 | ||
| GenBank mRNA | BT099811.1 | - | 2 - 3 | 217 | 73 | |
More Information
Set Membership: |
|

TpnC73F
Genomic Map