Assay Details
| Gene Symbol: Rbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104162.4 | NP_001097632.1 | 5 - 6 | 1171 | 75 | |
| NM_001144489.3 | NP_001137961.1 | 5 - 6 | 1171 | 75 | ||
| NM_001259883.2 | NP_001246812.1 | 5 - 6 | 1171 | 75 | ||
| NM_001259884.2 | NP_001246813.1 | 5 - 6 | 1171 | 75 | ||
| NM_001275050.2 | NP_001261979.1 | 10 - 11 | 1650 | 75 | ||
| NM_168718.3 | NP_730245.2 | 10 - 11 | 1650 | 75 | ||
| GenBank mRNA | AY122226.1 | - | 10 - 11 | 1648 | 75 | |
More Information
Set Membership: |
|

Rbp6
Genomic Map