Assay Details
| Gene Symbol: GNBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079417.3 | NP_524141.1 | 2 - 3 | 628 | 58 | |
| NM_168771.2 | NP_730350.1 | 3 - 4 | 566 | 58 | ||
| NM_168772.2 | NP_730351.1 | 2 - 3 | 468 | 58 | ||
| GenBank mRNA | AF228473.1 | - | 2 - 3 | 607 | 58 | |
| BT099533.1 | - | 2 - 3 | 642 | 58 | ||
More Information
Set Membership: |
|

GNBP2
Genomic Map