Assay Details
| Gene Symbol: Indy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169994.2 | NP_001163465.1 | 6 - 7 | 863 | 96 | |
| NM_079426.4 | NP_524150.1 | 6 - 7 | 766 | 96 | ||
| NM_168778.2 | NP_730363.2 | 6 - 7 | 854 | 96 | ||
| NM_168779.2 | NP_730364.1 | 6 - 7 | 882 | 96 | ||
| GenBank mRNA | AF509505.1 | - | 6 - 7 | 849 | 96 | |
| AY102686.1 | - | 6 - 7 | 730 | 96 | ||
| BT100230.1 | - | 6 - 7 | 848 | 96 | ||
More Information
Set Membership: |
|

Indy
Genomic Map