Assay Details
| Gene Symbol: Indy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169994.2 | NP_001163465.1 | 7 - 8 | 1316 | 95 | |
| NM_079426.4 | NP_524150.1 | 7 - 8 | 1219 | 95 | ||
| NM_168778.2 | NP_730363.2 | 7 - 8 | 1307 | 95 | ||
| NM_168779.2 | NP_730364.1 | 7 - 8 | 1335 | 95 | ||
| GenBank mRNA | AF509505.1 | - | 7 - 8 | 1302 | 95 | |
| AY102686.1 | - | 7 - 8 | 1183 | 95 | ||
| BT100230.1 | - | 7 - 8 | 1301 | 95 | ||
More Information
Set Membership: |
|

Indy
Genomic Map