Assay Details
Gene Symbol: nes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079433.3 | NP_524157.2 | 3 - 4 | 1293 | 125 | |
NM_168788.3 | NP_730390.1 | 3 - 4 | 1180 | 125 | ||
NM_176349.2 | NP_788527.1 | 3 - 4 | 1277 | 125 | ||
GenBank mRNA | AY070936.1 | - | 3 - 4 | 1176 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
