Assay Details
Gene Symbol: pip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079434.3 | NP_524158.2 | 2 - 3 | 436 | 64 | |
NM_168793.2 | NP_730402.1 | 2 - 3 | 436 | 64 | ||
NM_176351.4 | NP_788529.1 | 2 - 3 | 436 | 64 | ||
NM_176352.2 | NP_788530.1 | 2 - 3 | 436 | 64 | ||
NM_176353.2 | NP_788531.1 | 2 - 3 | 436 | 64 | ||
NM_176354.2 | NP_788532.1 | 2 - 3 | 436 | 64 | ||
NM_176355.2 | NP_788533.1 | 2 - 3 | 436 | 64 | ||
NM_176356.2 | NP_788534.1 | 2 - 3 | 436 | 64 | ||
NM_176357.2 | NP_788535.2 | 2 - 3 | 436 | 64 | ||
NM_176358.2 | NP_788536.1 | 2 - 3 | 436 | 64 | ||
NM_176359.2 | NP_788537.1 | 2 - 3 | 436 | 64 | ||
GenBank mRNA | BT010278.1 | - | 2 - 3 | 426 | 64 | |
BT125816.1 | - | 2 - 3 | 426 | 64 | ||
BT125857.1 | - | 2 - 3 | 410 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
