Assay Details
Gene Symbol: Rcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140868.2 | NP_649125.1 | 1 - 2 | 355 | 94 | |
NM_168803.2 | NP_730429.2 | 2 - 3 | 716 | 94 | ||
GenBank mRNA | BT044243.1 | - | 2 - 3 | 716 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
