Assay Details
Gene Symbol: SREBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275135.1 | NP_001262064.1 | 3 - 4 | 1824 | 103 | |
NM_079442.3 | NP_524166.2 | 2 - 3 | 1722 | 103 | ||
NM_168815.2 | NP_730449.1 | 3 - 4 | 1745 | 103 | ||
NM_168816.2 | NP_730450.1 | 3 - 4 | 1855 | 103 | ||
GenBank mRNA | BT010285.1 | - | 3 - 4 | 1736 | 103 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
