Assay Details
Gene Symbol: Prp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140899.2 | NP_649156.1 | 1 - 2 | 1221 | 68 | |
NM_168825.3 | NP_730463.1 | 2 - 3 | 1155 | 68 | ||
GenBank mRNA | AY119640.1 | - | 2 - 3 | 1110 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -




Gene Ontology Categories:
