Assay Details
Gene Symbol: eRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275175.1 | NP_001262104.1 | 3 - 4 | 472 | 68 | |
NM_001275176.1 | NP_001262105.1 | 3 - 4 | 472 | 68 | ||
NM_140953.3 | NP_649210.1 | 2 - 3 | 1028 | 68 | ||
NM_168845.2 | NP_730517.1 | 3 - 4 | 508 | 68 | ||
NM_168846.2 | NP_730518.1 | 3 - 4 | 472 | 68 | ||
NM_168847.2 | NP_730519.1 | 3 - 4 | 737 | 68 | ||
NM_168848.2 | NP_730520.1 | 3 - 4 | 495 | 68 | ||
NM_176369.3 | NP_788547.1 | 2 - 3 | 446 | 68 | ||
GenBank mRNA | AY069511.1 | - | 2 - 3 | 473 | 68 | |
BT120009.1 | - | 3 - 4 | 479 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
