Assay Details
Gene Symbol: eRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275175.1 | NP_001262104.1 | 6 - 7 | 1296 | 80 | |
NM_001275176.1 | NP_001262105.1 | 6 - 7 | 1296 | 80 | ||
NM_140953.3 | NP_649210.1 | 5 - 6 | 1852 | 80 | ||
NM_168845.2 | NP_730517.1 | 6 - 7 | 1332 | 80 | ||
NM_168846.2 | NP_730518.1 | 6 - 7 | 1296 | 80 | ||
NM_168847.2 | NP_730519.1 | 6 - 7 | 1561 | 80 | ||
NM_168848.2 | NP_730520.1 | 6 - 7 | 1319 | 80 | ||
NM_176369.3 | NP_788547.1 | 5 - 6 | 1270 | 80 | ||
GenBank mRNA | AY069511.1 | - | 5 - 6 | 1297 | 80 | |
BT120009.1 | - | 6 - 7 | 1303 | 80 | ||
BT120010.1 | - | 6 - 7 | 1306 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
