Assay Details
Gene Symbol: eRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140953.3 | NP_649210.1 | 6 - 7 | 2001 | 60 | |
NM_168845.2 | NP_730517.1 | 7 - 8 | 1481 | 60 | ||
NM_168846.2 | NP_730518.1 | 7 - 8 | 1445 | 60 | ||
NM_168847.2 | NP_730519.1 | 7 - 8 | 1710 | 60 | ||
NM_168848.2 | NP_730520.1 | 7 - 8 | 1468 | 60 | ||
NM_176369.3 | NP_788547.1 | 6 - 7 | 1419 | 60 | ||
GenBank mRNA | AY069511.1 | - | 6 - 7 | 1446 | 60 | |
BT120009.1 | - | 7 - 8 | 1452 | 60 | ||
BT120010.1 | - | 7 - 8 | 1455 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
