Assay Details
| Gene Symbol: jing | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103729.2 | NP_001097199.1 | 5 - 6 | 4371 | 59 | |
| NM_001103730.2 | NP_001097200.1 | 5 - 6 | 5642 | 59 | ||
| NM_206027.3 | NP_995749.2 | 5 - 6 | 4062 | 59 | ||
| GenBank mRNA | AF285778.1 | - | 4 - 5 | 3835 | 59 | |
| BT025201.1 | - | 5 - 6 | 4042 | 59 | ||
More Information
Set Membership: |
|

jing
Genomic Map