Assay Details
| Gene Symbol: Eb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273805.1 | NP_001260734.1 | 2 - 3 | 228 | 79 | |
| NM_136389.5 | NP_610233.1 | 2 - 3 | 321 | 79 | ||
| NM_165487.4 | NP_724495.2 | 2 - 3 | 321 | 79 | ||
| NM_165488.3 | NP_724496.1 | 2 - 3 | 321 | 79 | ||
| NM_165489.3 | NP_724497.1 | 2 - 3 | 227 | 79 | ||
| NM_206030.3 | NP_995752.1 | 2 - 3 | 321 | 79 | ||
| GenBank mRNA | AF132560.1 | - | 2 - 3 | 314 | 79 | |
| AY071380.1 | - | 2 - 3 | 355 | 79 | ||
| BT100289.1 | - | 2 - 3 | 295 | 79 | ||
More Information
Set Membership: |
|

Eb1
Genomic Map