Assay Details
| Gene Symbol: mim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014500.3 | NP_001014500.2 | 12 - 13 | 5072 | 144 | |
| NM_001103735.3 | NP_001097205.1 | 12 - 13 | 5072 | 144 | ||
| NM_001201940.3 | NP_001188869.1 | 12 - 13 | 5072 | 144 | ||
| NM_001201941.2 | NP_001188870.1 | 12 - 13 | 5018 | 144 | ||
| NM_001259228.2 | NP_001246157.1 | 12 - 13 | 3518 | 144 | ||
| NM_001259229.2 | NP_001246158.1 | 13 - 14 | 5153 | 144 | ||
| NM_001273818.1 | NP_001260747.1 | 11 - 12 | 2714 | 144 | ||
More Information
Set Membership: |
|

mim
Genomic Map