Assay Details
Gene Symbol: brp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043070.1 | NP_001036535.1 | 13 - 14 | 4326 | 58 | |
NM_001103771.4 | NP_001097241.2 | 16 - 17 | 5227 | 58 | ||
NM_001259298.2 | NP_001246227.1 | 9 - 10 | 4354 | 58 | ||
NM_001259299.3 | NP_001246228.1 | 17 - 18 | 6583 | 58 | ||
NM_001259300.2 | NP_001246229.1 | 14 - 15 | 4990 | 58 | ||
NM_001259301.2 | NP_001246230.1 | 16 - 17 | 5212 | 58 | ||
NM_001273897.2 | NP_001260826.1 | 11 - 12 | 4110 | 58 | ||
NM_165677.4 | NP_724796.3 | 17 - 18 | 5471 | 58 | ||
NM_206063.3 | NP_995785.2 | 15 - 16 | 4836 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
