Assay Details
Gene Symbol: Mmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273901.1 | NP_001260830.1 | 3 - 4 | 929 | 54 | |
NM_136667.3 | NP_610511.3 | 4 - 5 | 1201 | 54 | ||
NM_206066.3 | NP_995788.1 | 5 - 6 | 1673 | 54 | ||
GenBank mRNA | AJ289232.1 | - | 6 - 7 | 1543 | 54 | |
BT150239.1 | - | 5 - 6 | 1637 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
