Assay Details
| Gene Symbol: Galphao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273940.1 | NP_001260869.1 | 3 - 4 | 457 | 67 | |
| NM_078960.5 | NP_523684.2 | 6 - 7 | 1102 | 67 | ||
| NM_165772.4 | NP_724934.1 | 6 - 7 | 984 | 67 | ||
| NM_165773.3 | NP_724935.1 | 7 - 8 | 1039 | 67 | ||
| NM_176124.3 | NP_788304.1 | 6 - 7 | 911 | 67 | ||
| NM_176125.3 | NP_788305.1 | 6 - 7 | 966 | 67 | ||
| NM_176126.3 | NP_788306.1 | 6 - 7 | 884 | 67 | ||
| NM_176127.3 | NP_788307.1 | 6 - 7 | 994 | 67 | ||
| NM_206079.2 | NP_995801.1 | 5 - 6 | 1114 | 67 | ||
| NM_206080.2 | NP_995802.1 | 5 - 6 | 866 | 67 | ||
| GenBank mRNA | AY121631.1 | - | 7 - 8 | 1020 | 67 | |
| BT046134.2 | - | 6 - 7 | 1119 | 67 | ||
| BT124767.1 | - | 3 - 4 | 457 | 67 | ||
| M29602.1 | - | 5 - 6 | 1879 | 67 | ||
| M29731.1 | - | 6 - 7 | 888 | 67 | ||
| M29732.1 | - | 6 - 7 | 1242 | 67 | ||
| M30151.1 | - | 6 - 7 | 912 | 67 | ||
| M86660.1 | - | 5 - 6 | 977 | 67 | ||
More Information
Set Membership: |
|

Galphao
Genomic Map