Assay Details
Gene Symbol: Galphao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273940.1 | NP_001260869.1 | 4 - 5 | 613 | 59 | |
NM_078960.5 | NP_523684.2 | 7 - 8 | 1258 | 59 | ||
NM_165772.4 | NP_724934.1 | 7 - 8 | 1140 | 59 | ||
NM_165773.3 | NP_724935.1 | 8 - 9 | 1195 | 59 | ||
NM_176124.3 | NP_788304.1 | 7 - 8 | 1067 | 59 | ||
NM_176125.3 | NP_788305.1 | 7 - 8 | 1122 | 59 | ||
NM_176126.3 | NP_788306.1 | 7 - 8 | 1040 | 59 | ||
NM_176127.3 | NP_788307.1 | 7 - 8 | 1150 | 59 | ||
NM_206079.2 | NP_995801.1 | 6 - 7 | 1270 | 59 | ||
NM_206080.2 | NP_995802.1 | 6 - 7 | 1022 | 59 | ||
GenBank mRNA | AY121631.1 | - | 8 - 9 | 1176 | 59 | |
BT046134.2 | - | 7 - 8 | 1275 | 59 | ||
BT124767.1 | - | 4 - 5 | 613 | 59 | ||
M29602.1 | - | 6 - 7 | 2035 | 59 | ||
M29731.1 | - | 7 - 8 | 1044 | 59 | ||
M29732.1 | - | 7 - 8 | 1398 | 59 | ||
M30151.1 | - | 7 - 8 | 1068 | 59 | ||
M86660.1 | - | 6 - 7 | 1133 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
