Assay Details
Gene Symbol: psq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014520.3 | NP_001014520.2 | 2 - 3 | 912 | 92 | |
NM_001316414.1 | NP_001303343.1 | 2 - 3 | 912 | 92 | ||
NM_001382049.1 | NP_001369078.1 | 2 - 3 | 912 | 92 | ||
NM_078962.4 | NP_523686.2 | 2 - 3 | 912 | 92 | ||
NM_165790.3 | NP_724955.1 | 2 - 3 | 912 | 92 | ||
NM_206086.3 | NP_995808.1 | 2 - 3 | 755 | 92 | ||
GenBank mRNA | AY069588.1 | - | 2 - 3 | 833 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
