Assay Details
Gene Symbol: psq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014518.3 | NP_001014518.1 | 5 - 6 | 2739 | 56 | |
NM_001014519.3 | NP_001014519.1 | 5 - 6 | 2988 | 56 | ||
NM_001014520.3 | NP_001014520.2 | 9 - 10 | 3418 | 56 | ||
NM_001316414.1 | NP_001303343.1 | 9 - 10 | 3418 | 56 | ||
NM_001382049.1 | NP_001369078.1 | 9 - 10 | 3418 | 56 | ||
NM_001382050.1 | NP_001369079.1 | 6 - 7 | 2824 | 56 | ||
NM_001382051.1 | NP_001369080.1 | 6 - 7 | 2824 | 56 | ||
NM_078962.4 | NP_523686.2 | 8 - 9 | 3466 | 56 | ||
NM_165790.3 | NP_724955.1 | 9 - 10 | 3412 | 56 | ||
NM_165791.3 | NP_724958.1 | 6 - 7 | 2824 | 56 | ||
NM_165792.3 | NP_724959.1 | 7 - 8 | 2563 | 56 | ||
NM_176141.3 | NP_788321.1 | 7 - 8 | 2879 | 56 | ||
NM_176142.3 | NP_788322.1 | 7 - 8 | 2612 | 56 | ||
NM_176143.3 | NP_788323.1 | 6 - 7 | 2649 | 56 | ||
NM_206086.3 | NP_995808.1 | 8 - 9 | 3309 | 56 | ||
GenBank mRNA | AY069588.1 | - | 8 - 9 | 3387 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
