Assay Details
Gene Symbol: casp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299544.1 | NP_001286473.1 | 8 - 9 | 1339 | 68 | |
NM_137236.5 | NP_611080.1 | 8 - 9 | 1354 | 68 | ||
NM_206126.2 | NP_995848.1 | 9 - 10 | 1354 | 68 | ||
GenBank mRNA | AY122148.1 | - | 8 - 9 | 1295 | 68 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
