Assay Details
Gene Symbol: EDTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079046.3 | NP_523770.2 | 3 - 4 | 2088 | 64 | |
GenBank mRNA | AB036800.2 | - | 3 - 4 | 2082 | 64 | |
AF145681.1 | - | 3 - 4 | 2066 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
