Assay Details
Gene Symbol: EDTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079046.3 | NP_523770.2 | 5 - 6 | 2591 | 69 | |
NM_206151.2 | NP_995873.2 | 5 - 6 | 2612 | 69 | ||
GenBank mRNA | AB036800.2 | - | 5 - 6 | 2585 | 69 | |
AF145681.1 | - | 5 - 6 | 2569 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
