Assay Details
Gene Symbol: mei-W68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206179.3 | NP_995901.1 | 2 - 3 | 1257 | 81 | |
GenBank mRNA | AF085276.1 | - | 2 - 3 | 1232 | 81 | |
BT120379.1 | - | 1 - 2 | 371 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
