Assay Details
| Gene Symbol: RIC-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103919.3 | NP_001097389.1 | 3 - 4 | 714 | 96 | |
| NM_001103921.3 | NP_001097391.1 | 3 - 4 | 669 | 96 | ||
| NM_001169750.2 | NP_001163221.1 | 3 - 4 | 714 | 96 | ||
| NM_001169755.2 | NP_001163226.1 | 3 - 4 | 714 | 96 | ||
| NM_001169756.2 | NP_001163227.1 | 3 - 4 | 714 | 96 | ||
| NM_001274184.2 | NP_001261113.1 | 3 - 4 | 572 | 96 | ||
| NM_001274185.2 | NP_001261114.1 | 3 - 4 | 714 | 96 | ||
| NM_001299729.1 | NP_001286658.1 | 3 - 4 | 565 | 96 | ||
| NM_001299730.1 | NP_001286659.1 | 3 - 4 | 714 | 96 | ||
| NM_137693.4 | NP_611537.2 | 3 - 4 | 714 | 96 | ||
| GenBank mRNA | BT029115.1 | - | 3 - 4 | 637 | 96 | |
More Information
Set Membership: |
|

RIC-3
Genomic Map