Assay Details
| Gene Symbol: RIC-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103919.3 | NP_001097389.1 | 8 - 9 | 1411 | 70 | |
| NM_001103921.3 | NP_001097391.1 | 7 - 8 | 1345 | 70 | ||
| NM_001169752.2 | NP_001163223.1 | 7 - 8 | 1291 | 70 | ||
| NM_001169754.2 | NP_001163225.1 | 7 - 8 | 1291 | 70 | ||
| NM_001169756.2 | NP_001163227.1 | 7 - 8 | 1390 | 70 | ||
| NM_001169759.2 | NP_001163230.1 | 6 - 7 | 1270 | 70 | ||
| NM_001274184.2 | NP_001261113.1 | 7 - 8 | 1248 | 70 | ||
| NM_137693.4 | NP_611537.2 | 7 - 8 | 1390 | 70 | ||
| NM_166418.4 | NP_726018.2 | 6 - 7 | 1270 | 70 | ||
| NM_206188.4 | NP_995910.2 | 7 - 8 | 1251 | 70 | ||
| GenBank mRNA | BT029115.1 | - | 8 - 9 | 1334 | 70 | |
More Information
Set Membership: |
|

RIC-3
Genomic Map