Assay Details
| Gene Symbol: RIC-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103921.3 | NP_001097391.1 | 8 - 9 | 1492 | 63 | |
| NM_001169750.2 | NP_001163221.1 | 8 - 9 | 1390 | 63 | ||
| NM_001169751.2 | NP_001163222.1 | 8 - 9 | 1291 | 63 | ||
| NM_001169752.2 | NP_001163223.1 | 8 - 9 | 1438 | 63 | ||
| NM_001169757.2 | NP_001163228.1 | 7 - 8 | 1270 | 63 | ||
| NM_001274184.2 | NP_001261113.1 | 8 - 9 | 1395 | 63 | ||
| NM_137693.4 | NP_611537.2 | 8 - 9 | 1537 | 63 | ||
| NM_166418.4 | NP_726018.2 | 7 - 8 | 1417 | 63 | ||
| NM_206188.4 | NP_995910.2 | 8 - 9 | 1398 | 63 | ||
More Information
Set Membership: |
|

RIC-3
Genomic Map