Assay Details
| Gene Symbol: MESK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079080.3 | NP_523804.3 | 4 - 5 | 778 | 97 | |
| NM_166454.2 | NP_726076.1 | 5 - 6 | 1206 | 97 | ||
| NM_166455.2 | NP_726077.1 | 5 - 6 | 1206 | 97 | ||
| NM_166456.3 | NP_726078.1 | 5 - 6 | 1206 | 97 | ||
| NM_206190.2 | NP_995912.1 | 5 - 6 | 1179 | 97 | ||
| NM_206191.2 | NP_995913.1 | 5 - 6 | 985 | 97 | ||
| NM_206192.2 | NP_995914.1 | 4 - 5 | 790 | 97 | ||
| NM_206193.2 | NP_995915.1 | 3 - 4 | 694 | 97 | ||
| NM_206194.2 | NP_995916.1 | 4 - 5 | 1122 | 97 | ||
| GenBank mRNA | AF195792.1 | - | 4 - 5 | 765 | 97 | |
| AY069037.1 | - | 5 - 6 | 1167 | 97 | ||
| BT001407.1 | - | 4 - 5 | 588 | 97 | ||
| BT001773.1 | - | 3 - 4 | 599 | 97 | ||
| BT003794.1 | - | 5 - 6 | 986 | 97 | ||
More Information
Set Membership: |
|

MESK2
Genomic Map