Assay Details
| Gene Symbol: Pal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299847.1 | NP_001286776.1 | 5 - 6 | 1140 | 70 | |
| NM_080379.5 | NP_525118.2 | 5 - 6 | 1028 | 70 | ||
| NM_206207.3 | NP_995929.2 | 5 - 6 | 1028 | 70 | ||
| GenBank mRNA | AY119165.1 | - | 5 - 6 | 1058 | 70 | |
More Information
Set Membership: |
|

Pal2
Genomic Map