Assay Details
Gene Symbol: Slik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166669.4 | NP_726441.1 | 13 - 14 | 5344 | 52 | |
NM_206213.3 | NP_995935.1 | 13 - 14 | 5737 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
