Assay Details
Gene Symbol: Slik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014549.3 | NP_001014549.1 | 3 - 4 | 706 | 61 | |
NM_001144280.2 | NP_001137752.1 | 3 - 4 | 665 | 61 | ||
NM_138064.4 | NP_611908.2 | 3 - 4 | 706 | 61 | ||
NM_166669.4 | NP_726441.1 | 3 - 4 | 706 | 61 | ||
NM_206213.3 | NP_995935.1 | 3 - 4 | 1099 | 61 | ||
NM_206214.4 | NP_995936.2 | 3 - 4 | 706 | 61 | ||
GenBank mRNA | BT150436.1 | - | 3 - 4 | 714 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
