Assay Details
Gene Symbol: Slik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014549.3 | NP_001014549.1 | 8 - 9 | 1788 | 114 | |
NM_001144280.2 | NP_001137752.1 | 8 - 9 | 1747 | 114 | ||
NM_138064.4 | NP_611908.2 | 8 - 9 | 1788 | 114 | ||
NM_166669.4 | NP_726441.1 | 8 - 9 | 1788 | 114 | ||
NM_206213.3 | NP_995935.1 | 8 - 9 | 2181 | 114 | ||
NM_206214.4 | NP_995936.2 | 9 - 10 | 1830 | 114 | ||
GenBank mRNA | BT150436.1 | - | 8 - 9 | 1796 | 114 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
