Assay Details
| Gene Symbol: E(bx) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169833.2 | NP_001163304.1 | 6 - 7 | 6930 | 63 | |
| NM_001169834.2 | NP_001163305.1 | 7 - 8 | 6503 | 63 | ||
| NM_001274258.1 | NP_001261187.1 | 6 - 7 | 6563 | 63 | ||
| NM_001274259.1 | NP_001261188.1 | 6 - 7 | 6975 | 63 | ||
| NM_001274260.1 | NP_001261189.1 | 6 - 7 | 6560 | 63 | ||
| NM_001274261.1 | NP_001261190.1 | 6 - 7 | 6472 | 63 | ||
| NM_167817.3 | NP_728505.1 | 7 - 8 | 6915 | 63 | ||
| NM_167819.3 | NP_728507.1 | 6 - 7 | 6975 | 63 | ||
| NM_206224.3 | NP_995946.1 | 6 - 7 | 6975 | 63 | ||
More Information
Set Membership: |
|

E(bx)
Genomic Map