Assay Details
Gene Symbol: nSyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259618.2 | NP_001246547.1 | 4 - 5 | 707 | 76 | |
NM_001274340.1 | NP_001261269.1 | 3 - 4 | 646 | 76 | ||
NM_001274341.1 | NP_001261270.1 | 3 - 4 | 658 | 76 | ||
NM_057710.6 | NP_477058.1 | 3 - 4 | 634 | 76 | ||
NM_167904.3 | NP_728644.1 | 3 - 4 | 637 | 76 | ||
NM_167905.4 | NP_728645.1 | 4 - 5 | 710 | 76 | ||
NM_167906.3 | NP_728646.2 | 4 - 5 | 707 | 76 | ||
NM_206234.4 | NP_995956.1 | 4 - 5 | 707 | 76 | ||
GenBank mRNA | AY069045.1 | - | 2 - 3 | 615 | 76 | |
BT099571.1 | - | 3 - 4 | 685 | 76 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
