Assay Details
Gene Symbol: msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274402.1 | NP_001261331.1 | 3 - 4 | 421 | 68 | |
NM_001274403.1 | NP_001261332.1 | 3 - 4 | 341 | 68 | ||
NM_001274404.1 | NP_001261333.1 | 3 - 4 | 478 | 68 | ||
NM_079940.5 | NP_524679.3 | 3 - 4 | 341 | 68 | ||
NM_206249.3 | NP_995971.2 | 3 - 4 | 341 | 68 | ||
NM_206250.3 | NP_995972.1 | 3 - 4 | 324 | 68 | ||
NM_206251.3 | NP_995973.1 | 3 - 4 | 335 | 68 | ||
NM_206252.3 | NP_995974.1 | 3 - 4 | 341 | 68 | ||
GenBank mRNA | BT031263.1 | - | 3 - 4 | 321 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
