Assay Details
Gene Symbol: Tip60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272304.1 | NP_001259233.1 | 2 - 3 | 780 | 61 | |
NM_001272305.1 | NP_001259234.1 | 2 - 3 | 287 | 61 | ||
NM_131923.4 | NP_572151.1 | 2 - 3 | 825 | 61 | ||
GenBank mRNA | AY051795.1 | - | 2 - 3 | 780 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
