Assay Details
| Gene Symbol: Proc-R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014723.2 | NP_001014723.1 | 3 - 4 | 873 | 111 | |
| NM_001014724.2 | NP_001014724.1 | 3 - 4 | 540 | 111 | ||
| NM_001297939.1 | NP_001284868.1 | 2 - 3 | 652 | 111 | ||
| NM_131955.2 | NP_572183.1 | 3 - 4 | 634 | 111 | ||
| NM_167020.2 | NP_726950.1 | 3 - 4 | 829 | 111 | ||
| GenBank mRNA | AY128481.1 | - | 3 - 4 | 632 | 111 | |
More Information
Set Membership: |
|

Proc-R
Genomic Map