Assay Details
Gene Symbol: Proc-R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014723.2 | NP_001014723.1 | 5 - 6 | 1161 | 60 | |
NM_001014724.2 | NP_001014724.1 | 5 - 6 | 828 | 60 | ||
NM_001297939.1 | NP_001284868.1 | 4 - 5 | 940 | 60 | ||
NM_131955.2 | NP_572183.1 | 5 - 6 | 922 | 60 | ||
NM_167020.2 | NP_726950.1 | 5 - 6 | 1117 | 60 | ||
GenBank mRNA | AY128481.1 | - | 5 - 6 | 920 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
