Assay Details
| Gene Symbol: Proc-R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014723.2 | NP_001014723.1 | 6 - 7 | 1448 | 83 | |
| NM_001014724.2 | NP_001014724.1 | 6 - 7 | 1115 | 83 | ||
| NM_001297939.1 | NP_001284868.1 | 5 - 6 | 1227 | 83 | ||
| NM_131955.2 | NP_572183.1 | 6 - 7 | 1209 | 83 | ||
| NM_167020.2 | NP_726950.1 | 6 - 7 | 1404 | 83 | ||
| GenBank mRNA | AY128481.1 | - | 6 - 7 | 1207 | 83 | |
More Information
Set Membership: |
|

Proc-R
Genomic Map