Assay Details
| Gene Symbol: SPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297963.1 | NP_001284892.1 | 3 - 4 | 736 | 70 | |
| NM_001297964.1 | NP_001284893.1 | 3 - 4 | 817 | 70 | ||
| NM_001382034.1 | NP_001368972.1 | 4 - 5 | 1025 | 70 | ||
| NM_131997.2 | NP_572225.1 | 4 - 5 | 1025 | 70 | ||
| GenBank mRNA | AY095526.1 | - | 4 - 5 | 1020 | 70 | |
| EU443243.2 | - | 3 - 4 | 818 | 70 | ||
More Information
Set Membership: |
|

SPR
Genomic Map