Assay Details
Gene Symbol: Usp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297983.1 | NP_001284912.1 | 1 - 2 | 249 | 59 | |
NM_132046.3 | NP_572274.1 | 1 - 2 | 418 | 59 | ||
GenBank mRNA | AY069649.1 | - | 1 - 2 | 225 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
