Assay Details
| Gene Symbol: Ca-alpha1T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258615.1 | NP_001245544.1 | 16 - 17 | 4612 | 97 | |
| NM_132068.3 | NP_572296.2 | 17 - 18 | 4636 | 97 | ||
More Information
Set Membership: |
|

Ca-alpha1T
Genomic Map