Assay Details
| Gene Symbol: Pde11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103707.2 | NP_001097177.1 | 6 - 7 | 2299 | 120 | |
| NM_001201908.2 | NP_001188837.1 | 6 - 7 | 1993 | 120 | ||
| NM_001201909.2 | NP_001188838.1 | 6 - 7 | 2299 | 120 | ||
| NM_001299115.1 | NP_001286044.1 | 6 - 7 | 2166 | 120 | ||
| NM_001299116.1 | NP_001286045.1 | 7 - 8 | 1386 | 120 | ||
| NM_136041.2 | NP_609885.2 | 6 - 7 | 2052 | 120 | ||
More Information
Set Membership: |
|

Pde11
Genomic Map