Assay Details
| Gene Symbol: ACXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274391.1 | NP_001261320.1 | 7 - 8 | 2445 | 100 | |
| NM_138984.4 | NP_620469.2 | 7 - 8 | 2439 | 100 | ||
| GenBank mRNA | BT053678.1 | - | 7 - 8 | 2437 | 100 | |
More Information
Set Membership: |
|

ACXD
Genomic Map