Assay Details
| Gene Symbol: ACXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274391.1 | NP_001261320.1 | 8 - 9 | 2769 | 63 | |
| NM_138984.4 | NP_620469.2 | 8 - 9 | 2763 | 63 | ||
| GenBank mRNA | BT053678.1 | - | 8 - 9 | 2761 | 63 | |
More Information
Set Membership: |
|

ACXD
Genomic Map