Assay Details
Gene Symbol: ACXB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138989.3 | NP_620474.2 | 8 - 9 | 2530 | 85 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
