Assay Details
| Gene Symbol: ACXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259082.1 | NP_001246011.1 | 11 - 12 | 2851 | 98 | |
| NM_001259083.1 | NP_001246012.1 | 11 - 12 | 2861 | 98 | ||
| NM_001273494.1 | NP_001260423.1 | 11 - 12 | 2879 | 98 | ||
| NM_001298954.1 | NP_001285883.1 | 11 - 12 | 2812 | 98 | ||
| NM_138990.2 | NP_620475.2 | 11 - 12 | 2866 | 98 | ||
More Information
Set Membership: |
|

ACXA
Genomic Map